Learn More
E. coli serotype O/K, FITC, Polyclonal, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA128977
Description
Escherichia coli (E. coli) are Gram negative enteric bacteria that live in the human gut. Its presence in nature is an indication of human feacal pollution. E. coli also causes human disease such as urinary tract infections and neonatal meningitis. Infection with particular enterotoxigenic strains can lead to life threatening intestinal diseases. E. coli are distinguished immunologically by serotyping based on three types of antigen: the somatic or O antigen which corresponds to terminal sugars on the cell surface lipopolysaccharide, the capsular or K antigens and the flagellar or H antigens.
PA1-28977 detects E. coli O+K (FITC ) from bacteria samples.PA1-28977 has been successfully used in immunofluorescence applications. Acetone fixation of the antigen source is recommended prior to staining.The PA1-28977 immunogen is: Mixture of E. coli serotypes (E. coli).
Specifications
E. coli serotypes O + K | |
Polyclonal | |
FITC | |
Goat | |
IgG fraction | |
RUO | |
Bacteria | |
Antibody | |
IgG |
ELISA, Immunocytochemistry, Immunofluorescence | |
4.0mg/mL | |
Escherichia coli | |
Mixture of E. coli serotypes (E. coli). | |
100 μL | |
Primary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.