Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E-Selectin Rat anti-Human, Mouse, Clone: UZ4, MilliporeSigma™

Catalog No. MAB2153MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2153MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2153MI Supplier MilliporeSigma Supplier No. MAB2153
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-E-Selectin Antibody, lectin domain, clone UZ4 is an antibody against E-Selectin for use in ELISA, flow cytometry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E-Selectin
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone UZ4
Conjugate Unconjugated
Gene Accession No. NM_000450.1
Gene Symbols SELE, LECAM2, ESEL, ELAM, ELAM-1, CD62E, ELAM1
Host Species Rat
Immunogen IgM
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.