Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E1A (Ad2/Ad5) Mouse anti-Human, Clone: M73, MilliporeSigma™

Catalog No. 05599MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-599-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-599-MI Supplier MilliporeSigma Supplier No. 05599
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect E1A (Ad2/Ad5) with Anti-E1A (Ad2/Ad5) Antibody, clone M73 (Mouse Monoclonal Antibody), that has been shown to work in immunoprecipitation, western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E1A (Ad2/Ad5)
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone M73
Conjugate Unconjugated
Formulation 0.05M sodium phosphate, pH 7.4, 0.1% sodium azide and 0.2% gelatin
Gene Accession No. NP_040507
Gene Symbols E1A
Host Species Mouse
Immunogen trpE-Ad2 E1A fusion protein isolated from E. coli
Purification Method Protein A chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 4°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.