Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2A Mouse anti-Human, Unlabeled, Clone: G127-32, BD

Catalog No. BDB554077 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554077 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554077 Supplier BD Biosciences Supplier No. 554077
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B lymphoid cells go through many intermediate steps before becoming plasma cells which produce and secrete immunoglobulins (Ig). The earliest stage of B-cell differentiation is represented by pro-B lymphocytes that contain both the Ig heavy- and light-chain genes in the transcriptionally inactive germ line configuration. As pro-B lymphocytes differentiate into pre-B cells, Ig heavy-chain genes rearrange and are transcribed and translated. However, the Ig light-chain genes are not transcribed before the pre-B cells differentiate into mature Blymphocytes. The developmental regulation of the Ig gene expression is dependent on various sequences in the Ig enhancer region. One class of such regulatory sequence elements comprises the E-boxes which share the NNCANNTGNN consensus sequence. The E2 boxes are particularly interesting because they are present in muscle and pancreas-specific enhancers. A family of proteins binds to the E2 box. These proteins share a common amino acid sequence motif that is proposed to form two amphipathic helices interrupted by a loop, designated the helix-loop-helix (HLH) motif. The HLH motif mediates homo- as well as heterodimerization with other HLH proteins. Most HLH proteins possess a basic region located at the N terminal of the HLH region which is responsible for DNA binding. Two E2 box binding proteins have been described (E12 and E47) that arise as alternatively spliced form of the E2A gene. E12 and E47 are identical except in the HLH region. These two proteins have been directly implicated in the regulation of B-cell, muscle and pancreas-specific gene expression. Futhermore, studies have shown that relative levels of E47 are important for regulating the lineage of T cells in the thymus. For example, mice deficient in E47 demonstrate a change in the ratio of CD4/CD8 cells as well as a greater percentage of cells that stain for the mature CD4 marker, implicating E47 in selection of T cells. Clone G127-32 recognizes human E47. It cross-reacts with mouse E47. G127-32 does not recognize E12, the alternately spliced product of the E2A gene or E2-2, a closely reltated protein. Recombinant E47 was used as immunogen.

Host Species: Mouse
Clone: G127-32
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human E47
Formula Weight [Chemical]: 75kDa

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen E2A
Applications Western Blot
Classification Monoclonal
Clone G127-32
Concentration 0.5mg/mL
Conjugate Unconjugated
Description E47(E2A), Immunoglobulin E2-Box Binding Protein
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant Human E47
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.