Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F-1 Mouse anti-Human, Xenopus, Clone: KH20 & KH95, MilliporeSigma™

Catalog No. 05379MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-379-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-379-MI Supplier MilliporeSigma Supplier No. 05379
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-E2F-1 Antibody, clones KH20 and KH95 is a high quality Mouse Monoclonal Antibody for the detection of E2F-1 and has been validated in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F-1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone KH20 & KH95
Conjugate Unconjugated
Formulation Protein G Purified immunoglobulin in Protein A Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl and 0.05% sodium azide.
Gene Accession No. NM_005225
Gene Symbols E2F1, RBAP-1, E2F-1, RBP3, RBBP3, RBBP-3, PBR3
Host Species Mouse
Immunogen GST fusion protein corresponding to full-length human E2F-1.
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Xenopus species
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.