Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E2F4 Mouse anti-Human, Mouse, Clone: LLF4-2, MilliporeSigma™

Catalog No. MABE160MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE160MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE160MI Supplier MilliporeSigma Supplier No. MABE160
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-E2F4 Antibody, clone LLF4-2 is a Mouse Monoclonal Antibody for detection of E2F4 also known as Transcription factor E2F4, E2F-4 and has been validated in western blotting and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E2F4
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone LLF4-2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001941
Gene Symbols E2F4
Host Species Mouse
Immunogen Histidine-tagged recombinant protein corresponding to human E2F4.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.