Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E3 ubiquitin-protein ligase UHRF1 Mouse anti-Mouse, Clone: BF18.11, MilliporeSigma™

Catalog No. MABE977MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE977MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE977MI Supplier MilliporeSigma Supplier No. MABE977
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-E3 ubiquitin-protein ligase UHRF1, clone BF18.11 Antibody is validated for use in Western Blotting and Immunocytochemistry and Immunoprecipitation for the detection of E3 ubiquitin-protein ligase UHRF1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen E3 ubiquitin-protein ligase UHRF1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone BF18.11
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001104548
Gene Symbols Uhrf1, Np95
Host Species Mouse
Immunogen His-tagged recombinant protein corresponding to mouse E3 ubiquitin-protein ligase UHRF1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.