Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

E4F1, Mouse, Clone: 4B10, Abnova™

Catalog No. 89021382 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-382 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-382 Supplier Abnova Corporation Supplier No. H00001877M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant E4F1.

The zinc finger protein encoded by this gene is one of several cellular transcription factors whose DNA-binding activities are regulated through the action of adenovirus E1A. A 50kDa amino-terminal product is generated from the full-length protein through proteolytic cleavage. The protein is differentially regulated by E1A-induced phosphorylation. The full-length gene product represses transcription from the E4 promoter in the absence of E1A, while the 50kDa form acts as a transcriptional activator in its presence. [provided by RefSeq

Sequence: GHQIIVQNVTMDEETALGPEAAAADTITIATPESLTEQVAMTLASAISEGTVLAARAGTSGTEQATVTMVSSEDIEILEHAGELVIASPEGQLEVQTVIV

Specifications

Antigen E4F1
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 4B10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant E4F1.
Formulation PBS with no preservative; pH 7.4
Gene E4F1
Gene Accession No. NM_004424
Gene Alias E4F/MGC99614
Gene Symbols E4F1
Host Species Mouse
Immunogen E4F1 (NP_004415.2, 685 a.a. ∼ 784 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1877
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.