Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EAAT2 (Excitatory amino acid transporter 2; Glutamate transporter 1; GLT1) Mouse anti-Rat, Unlabeled, Clone: 20, BD

Catalog No. BDB611654 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611654 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611654 Supplier BD Biosciences Supplier No. 611654
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Electrogenic-coupled (Na+ and K+) glutamate transporters in nerve terminals and glial cells are essential for maintaining subtoxic levels of extracellular excitatory amino acid neurotransmitters (e.g. glutamate and aspartate). They are also important for modifying synaptic transmission through the cotransport of ions and neurotransmitters. Excitatory amino acid transporter 2 (EAAT2) has 8 α-helical transmembrane domains and two reentrant pore-loop-like structures whose conformation is altered during ion neurotransmitter transport. EAAT2 is primarily expressed in astrocytes in adult brain and spinal cord, however it is also found in axons during fetal development. Mice deficient in EAAT2 exhibit lethal epileptic seizures with destruction of hippocampal neurons and an increased susceptibility to cold-induced cortical injury. Such defects in glutamine transporters have also been implicated in stroke, brain trauma, Alzheimer's disease, amyotrophic lateral sclerosis, and Huntington's disease. Thus, glutamine transporters, such as EAAT2, are critical for prevention of neurotoxic brain injury and may regulate glutamatergic synaptic transmission.

Host Species: Mouse
Clone: 20
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Human EAAT2 aa. 562-574

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen EAAT2 (Excitatory amino acid transporter 2; Glutamate transporter 1; GLT1)
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Description Excitatory amino acid transporter 2; Glutamate transporter 1; GLT1
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human EAAT2 aa. 562-574
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.