Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Early B Lineage Rat anti-Mouse, FITC, Clone: AA4.1, BD

Catalog No. BDB559156 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559156 0.5 mg
BDB561990 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB559156 Supplier BD Biosciences Supplier No. 559156

Rat Monoclonal Antibody

The AA4.1 antibody reacts with a 130-140kDa type I transmembrane protein expressed on immature B lymphocytes in the adult bone marrow; and on hematopoietic progenitors and stem cells in adult bone marrow, fetal liver, and embryonic yolk sac. Although staining of splenic immature/transitional B cells has been reported, we find that the antigen density is much lower in the spleen than in the bone marrow. Staining of spleen requires amplification through the use of a second step. The FITC conjugate of mAb AA4.1, while ideal for bone marrow staining, is not effective in the spleen (please see Usage comments below). It has been observed that the staining pattern of mAb 493 (Mfr. No. 550433 and 550434 for the purified and biotinylated formats, respectively) is similar to that of mAb AA4.1, that both antibodies precipitate molecules of the same molecular weight, and that staining by mAb AA4.1 is not blocked by mAb 493, suggesting that the antibodies recognize separate epitopes of the same Early B Lineage antigen.

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Early B Lineage
Applications Flow Cytometry
Classification Monoclonal
Clone AA4.1
Concentration 0.5mg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Host Species Rat
Immunogen Pre-B lymphoma 70Z/3, derived from (C57BL/6 x DBA/2)F1 mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.