Learn More
Eastern Equine Encephalitis Mouse anti-Human, Clone: 1A4B.6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB8754
Description
Detect the Eastern Equine Encephalitis protein using this Anti-Eastern Equine Encephalitis, Virus, clone 1A4B.6 validated for use in ELISA, immunofluorescence and Hemagglutination Inhibition.
Specifications
Eastern Equine Encephalitis | |
Monoclonal | |
Unconjugated | |
Mouse | |
Primary | |
2-8°C in undiluted aliquots for up to 12 months. | |
IgG2b |
ELISA, Hemagglutination Assay, Immunofluorescence | |
1A4B.6 | |
Purified immunoglobulin. Liquid in PBS with 0.1% Sodium Azide. | |
100 μg | |
Human | |