Learn More
MilliporeSigma™ anti-Eastern Equine Encephalitis, Virus, Clone: 1A4B.6, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB8754/DEL
Description
Enzyme Immunoassay, Immunofluorescence
Specifications
Eastern Equine Encephalitis, Virus | |
Unconjugated | |
100 μg | |
Primary | |
IgG2b |
Monoclonal | |
Mouse | |
The MAb 1A4B-6 is an IgG2b antibody that reacts with an E1 epitope shared by all alphaviruses. | |