Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EB1 Mouse, Unlabeled, Clone: 5, BD

Catalog No. BDB610535 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610535 150 μg
BDB610534 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610535 Supplier BD Biosciences Supplier No. 610535
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Familial adenomatous polyposis is an autosomal inherited disease characterized by mutations in the APC (adenomatous polyposis coli) gene. Individuals with this disease are predisposed to colorectal polyps and cancer. It has been proposed that disruption of the APC gene plays a role in the disease. APC is a large coiled-coil protein that interacts, via its carboxy terminus, with catenins and with the protein, EB1. This 30kDa protein was discovered as a result of its strong binding to the C-terminal domain of APC. Like the APC-binding proteins β-Catenin and pp120Cas, EB1 may play an essential role in modulating APC activity and tumorigenesis.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen EB1
Applications Western Blot
Classification Monoclonal
Clone 5
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.