Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBF1 (M01J), Mouse anti-Human, Clone: 1C12, Abnova™

Catalog No. 89021383 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-383 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-383 Supplier Abnova Corporation Supplier No. H00001879M01J

Mouse monoclonal antibody raised against a full-length recombinant EBF1.

Sequence: MFGIQESIQRSGSSMKEEPLGSGMNAVRTWMQGAGVLDANTAAQSGVGLARAHFEKQPPSNLRKSNFFHFVLALYDRQGQPVEIERTAFVGFVEKEKEANSEKTNNGIHYRLQLLYSNGIRTEQDFYVRLIDSMTKQAIVYEGQDKNPEMCRVLLTHEIMCSRCCDKKSCGNRNETPSDPVIIDRFFSKFFLKCNQNCLKNAGNPRDMRRFQVVVSTTVNVDGHVLAVSDNMFVHNNSKHGRRARRLDPSEGTPSYLEHATPCIKAISPSEGWTTGGATVIIIGDNFFDGLQVIFGTMLVWSELITPHAIRVQTPPRHIPGVVEVTLSYKSKQFCKGTPGRFIYTALNEPTIDYGFQRLQKVIPRHPGDPERLPKEVILKRAADLVEALYGMPHNNQEIILKRAADIAEALYSVPRNHNQLPALANTSVHAGMMGVNSFSGQLAVNVSEASQATNQGFTRNSSSVSPHGYVPSTTPQQTNYNSVTTSMNGYGSAAMSNLGGSPTFLNGSAANSPYAIVPSSPTMASSTSLPSNCSSSSGIFSFSPANMVSAVKQKSAFAPVVRPQTSPPPTCTSTNGNSLQAISGMIVPPM

Specifications

Antigen EBF1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant EBF1.
Formulation PBS with no preservative; pH 7.4
Gene EBF1
Gene Accession No. BC038805
Gene Alias COE1/EBF/FLJ39389/FLJ41763/O/E-1/OLF1
Gene Symbols EBF1
Host Species Mouse
Immunogen EBF1 (AAH38805, 1 a.a. ∼ 591 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1879
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.