Learn More
EBI3 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT338
Description
Detect IL-27 using this rabbit polyclonal antibody, Anti-EBI3 Antibody validated for use in western blotting, immunohistochemistry and Immunofluorescence.
Specifications
EBI3 | |
Polyclonal | |
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide. | |
Rabbit | |
Affinity Purified | |
Primary | |
2-8°C for 12 months |
Immunofluorescence, Immunohistochemistry, Western Blot | |
Unconjugated | |
EBI3; IL27B | |
KLH-conjugated linear peptide corresponding to the C-terminus of human EBI3. | |
100 μg | |
Human |