Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-EBNA Clone: E8.26,

Catalog No. DP15L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
DP15L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DP15L100UG Supplier MilliporeSigma™ Supplier No. DP15L100UG

Mouse Monoclonal Antibody

Specifically detects EBNA Clone: E8.26 in EBV-Positive Cells samples, and it is validated for Immunofluorescence, Immunoprecipitation

EBNA, Epstein Barr Virus Nuclear Antigen, is viral encoded and present in all EBV-infected cells. The genomes of different EBV isolates are distinguished by characteristic deletions and restriction polymorphisms. The function of this protein is still unknown. Some potential functions include a role in the replication of the episomal viral DNA, regulation of viral gene expression, and involvement in the growth transformation of B lymphocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBNA
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone E8.26
Conjugate Unconjugated
Formulation Lyophilized from 20mM ammoinum bicarbonate, 100μg BSA.
Host Species Mouse
Immunogen EBNA protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽88kDa wild-type EBNA from EBV-transformed human cells as well as the ∽31.5kDa mutant form of EBNA in Raji cells.
Target Species Epstein-Barr virus (EBV)
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.