Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBNA2 Rat anti-Human, Clone: R3, MilliporeSigma™

Catalog No. MABE8MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE8MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE8MI Supplier MilliporeSigma Supplier No. MABE8
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-EBNA2 Antibody, clone R3 is a Rat Monoclonal Antibody for detection of EBNA2 also known as Epstein-Barr nuclear antigen 2 and has been validated in western blot, immunoprecipitation and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBNA2
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone R3
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. YP_001129441
Gene Symbols EBNA2
Host Species Rat
Immunogen GST-tagged recombinant protein corresponding to human EBNA2.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.