Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EBV MA-gp350/250 Mouse anti-Human, Clone: 2L10, MilliporeSigma™

Catalog No. MAB8183MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8183MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8183MI Supplier MilliporeSigma Supplier No. MAB8183
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect EBV MA-gp350/250 using this Anti-EBV MA-gp350/250 Antibody, clone 2L10 validated for use in ELISA, immunofluorescence and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EBV MA-gp350/250
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2L10
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M phosphate buffer, pH 7.6, 0.25M NaCl, with 0.1% sodium azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.