Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECA39 Mouse anti-Human, Unlabeled, Clone: 51, BD

Catalog No. BDB611271 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611271 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611271 Supplier BD Biosciences Supplier No. 611271
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The c-myc oncogene is important in proliferation, differentiation, apoptosis, and malignant transformation. Products of myc oncogenes are transcription factors that bind to the DNA sequence, CACGTG, and regulate the expression of multiple target genes. Several myc target genes include ECA39, p53, ornithine decarboxylase (ODC), alpha-prothymosin, and Cdc25A. There is a high degree of identity among the mouse, human, and yeast ECA39 proteins. The myc recognition binding site of ECA39 is located 3′ to the start site of transcription in the mouse and human genes, but this element is absent in the nematode and yeast. Additionally, the tissue specific expression of ECA39 during embryogenesis is similar between human and mouse. Disruption of the ECA39 gene in yeast results in an increased growth rate in comparison to wild type, such that G1 is shorter. Furthermore, ECA39 is expressed in c-myc induced tumors and displays significant homology to the prokaryotic branched-chain amino acid aminotransferase (BCAT). Thus, ECA39 may be involved in the regulation of the cell cycle, possibly at the G1 to S phase transition.

Host Species: Mouse
Clone: 51
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human ECA39 aa. 213-335
Formula Weight [Chemical]: 43kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen ECA39
Applications Western Blot
Classification Monoclonal
Clone 51
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human ECA39 aa. 213-335
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.