Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECE2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA535266
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
Specifications
| ECE2 | |
| Polyclonal | |
| PBS with 0.09% sodium azide | |
| O60344 | |
| ECE2 | |
| KLH conjugated synthetic peptide between 551-579 amino acids from the central region of human ECE2 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| ECE2 | |
| ECE2;KIAA0604; Endothelin-converting enzyme 2; Methyltransferase-like region; Endothelin-converting enzyme 2 region | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9718 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction