Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-EDA2R, Clone: 2A10, Abnova™

Catalog No. 89300293 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-293 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-293 Supplier Abnova Corporation Supplier No. H00060401M11
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen EDA2R
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EDA2R.
Formulation PBS with no preservative; pH 7.4
Gene EDA2R
Gene Accession No. BC034919.1
Gene Alias EDA-A2R/EDAA2R/TNFRSF27/XEDAR
Gene Symbols EDA2R
Host Species Mouse
Immunogen EDA2R (AAH34919.1, 2 a.a. ∼ 135 a.a) partial recombinant protein with mouse IgG2a-Fc tag.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60401
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.