Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDG1, Mouse, Clone: 1F11, Abnova™

Catalog No. 89021386 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-386 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-386 Supplier Abnova Corporation Supplier No. H00001901M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EDG1.

The protein encoded by this gene is structurally similar to G protein-coupled receptors and is highly expressed in endothelial cells. It binds the ligand sphingosine-1-phosphate with high affinity and high specificity, and suggested to be involved in the processes that regulate the differentiation of endothelial cells. Activation of this receptor induces cell-cell adhesion. [provided by RefSeq

Sequence: MGPTSVPLVKAHRSSVSDYVNYDIIVRHYNYTGKLNISADKENSIKL

Specifications

Antigen EDG1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1F11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EDG1.
Formulation PBS with no preservative; pH 7.4
Gene S1PR1
Gene Accession No. BC018650
Gene Alias CHEDG1/D1S3362/ECGF1/EDG-1/EDG1/FLJ58121/S1P1
Gene Symbols S1PR1
Host Species Mouse
Immunogen EDG1 (AAH18650, 1 a.a. ∼ 47 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1901
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.