Learn More
EDG3, Mouse, Clone: 2G11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant EDG3.
Supplier: Abnova Corporation H00001903M02
Description
This gene encodes a member of the EDG family of receptors, which are G protein-coupled receptors. This protein has been identified as a functional receptor for sphingosine 1-phosphate and likely contributes to the regulation of angiogenesis and vascular endothelial cell function. [provided by RefSeq
Sequence: SKEMRRAFFRLVCNCLVRGRGARASPIQPALDPSRSKSSSSNNSSHSPKVKEDLPHTAPSSCIMDKNAALQNGIFCNSpecifications
EDG3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_005226 | |
S1PR3 | |
EDG3 (NP_005217.2, 302 a.a. ∼ 378 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunohistochemistry (PFA fixed) | |
2G11 | |
Mouse monoclonal antibody raised against a partial recombinant EDG3. | |
S1PR3 | |
EDG-3/EDG3/FLJ37523/FLJ93220/LPB3/MGC71696/S1P3 | |
Mouse | |
Affinity chromatography | |
RUO | |
1903 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.