Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDN2, Mouse, Clone: 3B4-1C5, Abnova™

Catalog No. 89021389 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-021-389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-021-389 Supplier Abnova Corporation Supplier No. H00001907M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant EDN2.

This gene encodes a member of the endothelin protein family of secretory vasoconstrictive peptides. The preproprotein is processed to a short mature form which functions as a ligand for the endothelin receptors that initiate intracellular signaling events. This gene product is involved in a wide range of biological processes, such as hypertension and ovulation. [provided by RefSeq

Sequence: MVSVPTTWCSVALALLVALHEGKGQAAATLEQPASSSHAQGTHLRLRRCSCSSWLDKECVYFCHLDIIWVNTPEQTAPYGLGNPPRRRRRSLPRRCQCSSARDPACATFCLRRPWTEAGAVPSRKSPADVFQTGKTGATTGELLQRLRDISTVKSLFAKRQQEAMREPRSTHSRWRKR

Specifications

Antigen EDN2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3B4-1C5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant EDN2.
Formulation PBS with no preservative; pH 7.4
Gene EDN2
Gene Accession No. BC034393
Gene Alias ET2/PPET2
Gene Symbols EDN2
Host Species Mouse
Immunogen EDN2 (AAH34393, 1 a.a. ∼ 178 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1907
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.