Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

endothelin 3, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017586 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-586 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-586 Supplier Abnova Corporation Supplier No. H00001908D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human EDN3 protein.

The protein encoded by this gene is a member of the endothelin family. Endothelins are endothelium-derived vasoactive peptides involved in a variety of biological functions. The active form of this protein is a 21 amino acid peptide processed from the precursor protein. The active peptide is a ligand for endothelin receptor type B (EDNRB). The interaction of this endothelin with EDNRB is essential for development of neural crest-derived cell lineages, such as melanocytes and enteric neurons. Mutations in this gene and EDNRB have been associated with Hirschsprung disease (HSCR) and Waardenburg syndrome (WS), which are congenital disorders involving neural crest-derived cells. Four alternatively spliced transcript variants encoding three distinct isoforms have been observed. [provided by RefSeq

Sequence: MEPGLWLLFGLTVTSAAGFVPCSQSGDAGRRGVSQAPTAARSEGDCEETVAGPGEETVAGPGEGTVAPTALQGPSPGSPGQEQAAEGAPEHHRSRRCTCFTYKDKECVYYCHLDIIWINTPEQTVPYGLSNYRGSFRGKRSAGPLPGNLQLSHRPHLRCACVGRYDKACLHFCTQTLDVSSNSRTAEKTDKEEEGKVEVKDQQSKQALDLHHPKLMPGSGLALAPSTCPRCLFQEGAP

Specifications

Antigen endothelin 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human EDN3 protein.
Formulation PBS with no preservative; pH 7.4
Gene EDN3
Gene Accession No. NM_000114.2
Gene Alias ET3/MGC15067/MGC61498
Gene Symbols EDN3
Host Species Rabbit
Immunogen EDN3 (NP_000105.1, 1 a.a. ∼ 238 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1908
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.