Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EEA1 Mouse anti-Human, FITC, Clone: 14, BD

Catalog No. BDB612006 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612006 50 μg
BDB612007 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612006 Supplier BD Biosciences Supplier No. 612006

Mouse Monoclonal Antibody

Early endosomes are a major cytoplasmic sorting compartment from which receptors and ligands may be distributed to various sites within the cell. Early endosome antigen 1 (EEA1) is a 180kDa hydrophilic peripheral membrane protein present in cytosol and membrane fractions. Immunofluorescence studies show that EEA1 colocalizes to early endosomes with transferrin receptor and Rab5, but not with the late endosome-localizing Rab7. EEA1 is predominantly α-helical and shares 17-20% sequence identity with the myosins. It contains a calmodulin-binding IQ motif and metal-binding cysteine “finger” motifs. It is thought that EEA1 is required for vesicular transport of proteins through early endosomes and that these finger motifs are required for this activity.

Host Species: Mouse
Clone: 14
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Mouse TRE1 aa. 82-179

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen EEA1
Applications Immunofluorescence
Classification Monoclonal
Clone 14
Concentration 250μg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse TRE1 aa. 82-179
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.