Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EEA1 Rabbit anti-Human, Mouse, Polyclonal

Catalog No. 071820MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-182-0MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-182-0MI Supplier MilliporeSigma™ Supplier No. 071820
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-EEA1 Antibody is validated for use in western blot, IC for the detection of EEA1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EEA1
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols EEA1; ZFYVE2; MST105; MSTP105
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the C-terminus of human EEA1.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_003557
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.