Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EED Mouse anti-Human, Mouse, Clone: AA19, MilliporeSigma™

Catalog No. 051320MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-132-0MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-132-0MI Supplier MilliporeSigma Supplier No. 051320
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-EED Antibody, clone AA19 is a mouse monoclonal antibody for detection of EED also known as WD protein associating with integrin cytoplasmic tails 1 ,embryonic ectoderm development and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EED
Applications Western Blot
Classification Monoclonal
Clone AA19
Conjugate Unconjugated
Formulation Mouse monoclonal IgG2aκ ascites in buffer containing 0.1MTris-Glycine (pH7.4) 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_068676
Gene Symbols HEED , WAIT-1, WAIT1, hEED
Host Species Mouse
Immunogen Recombinant fusion protein of full length murine EED tagged with MBP.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Ascites
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.