Learn More
EEF1G, Mouse, Clone: S1, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant EEF1G.
Supplier: Abnova Corporation H00001937M03
Description
This gene encodes a subunit of the elongation factor-1 complex, which is responsible for the enzymatic delivery of aminoacyl tRNAs to the ribosome. This subunit contains an N-terminal glutathione transferase domain, which may be involved in regulating the assembly of multisubunit complexes containing this elongation factor and aminoacyl-tRNA synthetases. [provided by RefSeq
Sequence: MAAGTLYTYPENWRAFKALIAAQYSGAQVRVLSAPPHFHFGQTNRTPEFLRKFPAGKVPAFEGDDGFCVFESNAIAYHVSNEELRGSTPEAAAQVVQWVSFADSDIVPPASTWVFPTLGIMHHDKQATENAKEEVRRILGLLDAYLKTRTFLVGERVTLADITVVCTLLWLYKQVLEPSFRQAFPNTNRWFLTCINQPQFRAVLGEVKLCEKMAQFDAKKFAETQPKKDTPRKEKGSREEKQKPQAERKEEKKAAAPAPEEEMDECEQALAAEPKAKDPFAHLPKSTFVLDEFKRKYSNEDTLSVALPYFWEHFDKDGWSLWYSEYRFPEELTQTFMSCNLITGMFQRLDKLRKNAFASVILFGTNNSSSISGVWVFRGQELAFPLSPDWQVDYESYTWRKLDPGSEETQTLVREYFSWEGAFQHVGKAFNQGKIFKSpecifications
EEF1G | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC015813 | |
EEF1G | |
EEF1G (AAH15813.1, 1 a.a. ∼ 437 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunofluorescence, Immunoprecipitation, Western Blot | |
S1 | |
Mouse monoclonal antibody raised against a full-length recombinant EEF1G. | |
EEF1G | |
EF1G/GIG35 | |
Mouse | |
Affinity chromatography | |
RUO | |
1937 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.