Learn More
EEF2K, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human EEF2K peptide using ARM Technology.
Supplier: Abnova Corporation H00029904K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a highly conserved protein kinase in the calmodulin-mediated signaling pathway that links activation of cell surface receptors to cell division. This kinase is involved in the regulation of protein synthesis. It phosphorylates eukaryotic elongation factor 2 (EEF2) and thus inhibits the EEF2 function. The activity of this kinase is increased in many cancers and may be a valid target for anti-cancer treatment. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
EEF2K | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
EEF2K | |
EEF2K | |
A synthetic peptide of human EEF2K | |
RUO | |
29904 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
eukaryotic elongation factor-2 kinase | |
EEF2K | |
HSU93850, MGC45041, eEF-2K | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.