Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EF1α Mouse anti-Bacteria, Mammals, Yeast (S. cerevisiae), Clone: CBP-KK1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05235
Description
Detect EF1α using this Anti-EF1α Antibody, clone CBP-KK1 validated for use in immunoprecipitation and western blotting.
Specifications
EF1α | |
Monoclonal | |
Unconjugated | |
NM_006620 | |
Mouse | |
Protein G chromatography | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Immunoprecipitation, Western Blot | |
CBP-KK1 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl with 0.05% sodium azide. | |
EEF1A1, eEF1A-1, FLJ25721, EF-1-alpha-1, EEF-1, HNGC:16303, ALPHA, CCS-3, PTI1, MGC131894, EEF1A, CCS3, MGC102687, EF-Tu, MGC16224, GRAF-1EF, LENG7, EF1A | |
Crude calmodulin-binding proteins from Trypanosoma brucei isolated using calmodulin-affinity chromatography. Clone CBP-KK1. | |
200 μg | |
Bacteria, S. cerevisiae, Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction