Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EFNA3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89003263 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-003-263 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-003-263 Supplier Abnova Corporation Supplier No. H00001944A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant EFNA3.

This gene encodes a member of the ephrin (EPH) family. The ephrins and EPH-related receptors comprise the largest subfamily of receptor protein-tyrosine kinases and have been implicated in mediating developmental events, especially in the nervous system and in erythropoiesis. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. This gene encodes an EFNA class ephrin. [provided by RefSeq

Sequence: RREGYTVQVNVNDYLDIYCPHYNSSGVGPGAGPGPGGGAEQYVLYMVSRNGYRTCNASQGFKRWECNRPHAPHSPIKFSEKFQRYSAFSLGYEFHAGHEYY

Specifications

Antigen EFNA3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant EFNA3.
Formulation 50% glycerol
Gene EFNA3
Gene Accession No. NM_004952
Gene Alias EFL2/EPLG3/Ehk1-L/LERK3
Gene Symbols EFNA3
Host Species Mouse
Immunogen EFNA3 (NP_004943, 45 a.a. ∼ 145 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1944
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.