Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eg5 Mouse anti-Human, Unlabeled, Clone: 20, BD

Catalog No. BDB611187 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611187 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611187 Supplier BD Biosciences Supplier No. 611187

Mouse Monoclonal Antibody

The mitotic spindle apparatus equally distributes duplicated chromosomes to daughter cells. This process is mediated by the dynamic instability of the spindle microtubules and the forces generated by the action of cytoplasmic dynein and kinesin-related motor proteins (KRPs), such as CHO1/MKLP1. These motors hydrolyze ATP as they move progressively along the microtubules. They organize the microtubules into bipolar spindles and may also serve as bridges between microtubules and chromosome kinetochores or centrosomes. Eg5, identified in Xenopus laevis, is a plus-end directed KRP, associated with the mitotic spindle. Vertebrate Eg5 is a member of the KRP subfamily bimC. These family members share a high degree of sequence homology (>80% within the N-terminal motor domain) and may execute similiar functions. During mitosis, Eg5 is specifically phosphorylated at Thr-297, an evolutionarily conserved cdc2 phosphorylation site, by p34[cdc2]/cyclin B. Inhibition of phosphorylation blocks the interaction of Eg5 with centrosomes. Thus, Eg5 is a mitotic motor protein that regulates spindle formation in a phosphorylation-dependent manner.

Host Species: Mouse
Clone: 20
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Eg5 aa. 324-532
Formula Weight [Chemical]: 120kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Eg5
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Eg5 aa. 324-532
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.