Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

epidermal growth factor (beta-urogastrone), Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89018814 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-814 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-814 Supplier Abnova Corporation Supplier No. H00001950A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant EGF.

Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. [provided by RefSeq

Sequence: NASCTNTEGGYTCMCAGRLSEPGLICPDSTPPPHLREDDHHYSVRNSDSECPLSHDGYCLHDGVCMYIEALDKYACNCVVGYIGERCQYRDLKWWKLRHA

Specifications

Antigen epidermal growth factor (beta-urogastrone)
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant EGF.
Formulation 50% glycerol
Gene EGF
Gene Accession No. NM_001963
Gene Alias HOMG4/URG
Gene Symbols EGF
Host Species Mouse
Immunogen EGF (NP_001954, 926 a.a. ∼ 1025 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 1950
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.