Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-EGF, Polyclonal,

Catalog No. 071432MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-143-2MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-143-2MI Supplier MilliporeSigma™ Supplier No. 071432

Specifications

Antigen EGF
Applications Immunohistochemistry (Paraffin), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal IgG neat antiserum in buffer containing 0.02% sodium azide.
Gene Symbols EGF; URG
Host Species Rabbit
Immunogen Recombinant human EGF.
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001963.2
Target Species Human
Content And Storage -20°C to -80°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.