Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGF Receptor Mouse, FITC, Clone: 13, BD

Catalog No. BDB612554 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612554 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612554 Supplier BD Biosciences Supplier No. 612554

Mouse Monoclonal Antibody

Epidermal Growth Factor (EGF) elicits a variety of cellular responses that are initiated by EGF receptor (EGF-R) binding and activation of intrinsic tyrosine kinase activity. Following ligand binding, EGF receptor is autophosphorylated and, in turn, phosphorylates several other endogenous proteins including phospholipase Cγ. These events initiate a number of intracellular responses that include increased levels of intracellular Ca2+ and transient expression of the nuclear oncogene products c-Myc and c-Fos.

Immunofluorescence, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen EGF Receptor
Applications Immunofluorescence
Classification Monoclonal
Clone 13
Concentration 250μg/mL
Conjugate FITC
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human EGF Receptor aa. 1020-1046
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.