Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EGF Receptor Mouse, Unlabeled, Clone: 13, BD
Mouse Monoclonal Antibody
$287.50 - $568.50
Specifications
| Antigen | EGF Receptor |
|---|---|
| Clone | 13 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Epidermal Growth Factor (EGF) elicits a variety of cellular responses that are initiated by EGF receptor (EGF-R) binding and activation of intrinsic tyrosine kinase activity. Following ligand binding, EGF receptor is autophosphorylated and, in turn, phosphorylates several other endogenous proteins including phospholipase C γ . These events initiate a number of intracellular responses that include increased levels of intracellular Ca2+ and transient expression of the nuclear oncogene products c-Myc and c-Fos.Specifications
| EGF Receptor | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 13 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| Human EGF Receptor aa. 1020 to 1046 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title