Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-EGFR Clone: 528,

Catalog No. GR01L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
GR01L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GR01L100UG Supplier MilliporeSigma™ Supplier No. GR01L100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects EGFR Clone: 528 in Human samples, and it is validated for Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization

Epidermal growth factor (EGF) initiates its effects on cell growth through interaction with a 170,000 dalton cell surface glycoprotein receptor. Binding of EGF or transforming growth factor a to the receptor activates a tyrosine-specific protein kinase intrinsic to the EGF-receptor protein. The phosphotyrosine content of target cells increases rapidly and the EGF-receptor is phosphorylated on tyrosine residues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 528
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin, pH 7.5.
Host Species Mouse
Immunogen partially purified, human EGFR from A431 cells
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽170kDa EGF receptor protein in A431 cells and normal skin.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.