Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ EGFR Mouse anti-Human, Clone: 528

Catalog No. MABF119MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF119MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF119MI Supplier MilliporeSigma™ Supplier No. MABF119
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-EGFR Antibody, clone 528 (Azide-free) detects level of EGFR and has been published and validated for use in Immunoprecipitation, Neutralizing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 528
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2aκ in buffer containing PBS without preservatives.
Gene Accession No. NP_005219
Gene Symbols EGFR, ERBB, ERBB1, HER1
Host Species Mouse
Immunogen Partially purified human EGFR from A431 cells
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.