Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-EGFR Clone: EGFR.1,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ GR15L100UG
Description
Specifically detects EGFR Clone: EGFR.1 in Human samples, and it is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization
Epidermal growth factor (EGF) initiates its effects on cell growth through interaction with a 170,000 dalton cell surface glycoprotein receptor. Binding of EGF or transforming growth factor alpha to the receptor activates a tyrosine-specific protein kinase intrinsic to the EGF-receptor protein. The phosphotyrosine content of target cells increases rapidly and the EGF-receptor is phosphorylated on tyrosine residues.
Specifications
EGFR | |
Monoclonal | |
Unconjugated | |
Mouse | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, Immunofluorescence, Immunoprecipitation, Neutralization | |
EGFR.1 | |
In 50mM sodium phosphate, 0.2% gelatin, pH 7.5. | |
partially purified, human EGFR from A431 cells | |
RUO | |
Recognizes the ∽170kDa EGF receptor protein in A431 cells. For neutralization studies Cat. No. GR15L is recommended. | |
2°C to 8°C | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction