Learn More
anti-EGFR, Clone: ICR10, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Supplier: Abcam AB231100UG

Description
Host Species: Rat
Clone: ICR10
Isotype: IgG2a
Cross Reactivity: Mouse, Human
Immunogen: Extracellular domain of Human EGF-receptor from head and neck carcinoma
Applications:
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation
Specifications
EGFR | |
ICR10 | |
Unconjugated | |
Extracellular domain of Human EGF-receptor from head and neck carcinoma | |
Cancer and Tumor Biology, Signal Transduction | |
Human, Mouse |
Monoclonal | |
1mg/mL | |
Rat | |
100 μg | |
Primary | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.