Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR Mouse anti-Human, Clone: LA1, MilliporeSigma™

Catalog No. 05101MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-101-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-101-MI Supplier MilliporeSigma Supplier No. 05101
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect EGFR using this Anti-EGFR Antibody, neutralizing, clone LA1 validated for use in immunocytochemistry, immunoprecipitation, NEUT and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR
Applications Immunocytochemistry, Immunoprecipitation, Neutralization, Western Blot
Classification Monoclonal
Clone LA1
Conjugate Unconjugated
Formulation Lyophilized protein G Purified mouse immunoglobulin. Lyophilized from 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing no preservatives.
Gene Accession No. NM_005228.3; NM_201282.1; NM_201283.1; NM_201284.1
Gene Symbols EGFR, ERBB1, ERBB, mENA
Host Species Mouse
Immunogen Human A431 membrane proteins.
Purification Method Protein G purfied
Quantity 250 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Lyophilized: 2 years at −20°C; Rehydrated: 1 month at 4°C or 6 months at −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.