Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EGFR (cytoplasmic domain) Mouse anti-Human, Mouse, Rat, Clone: 8G6.2, MilliporeSigma™

Catalog No. 051047MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-104-7MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-104-7MI Supplier MilliporeSigma Supplier No. 051047
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect EGFR (cytoplasmic domain) using this Anti-EGFR (cytoplasmic domain) Antibody, clone 8G6.2 validated for use in immunocytochemistry, immunohistochemistry, immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EGFR (cytoplasmic domain)
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 8G6.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_005219
Gene Symbols EGFR, ERBB1, ERBB, mENA
Host Species Mouse
Immunogen Recombinant protein encoding the cytoplasmic domain (C-terminal) of human EGF Receptor.
Purification Method Protein G purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.