Learn More
EGFR Rabbit anti-Human, Mouse, Rat, Hamster, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | EGFR |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4. Store at 2-8°C. |
| Gene Symbols | EGFR; ERBB1; ERBB; mENA |
| Host Species | Rabbit |
| Immunogen | Ovalbumin conjugated synthetic peptide (ETKPNGIFKGPTAENAEYLRVAPPSSEFIGA) corresponding to amino acids 1156-1186 of the processed mouse EGF receptor (EGFR) C-terminal domain (Accession number Q01279). The immunizing sequence shares 28/31 identity with the |
| Purification Method | Protein A chromatography |
| Quantity | 200 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.