Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF-4E Mouse, Unlabeled, Clone: 87, BD

Catalog No. BDB610269 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610269 50 μg
BDB610270 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610269 Supplier BD Biosciences Supplier No. 610269
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The eukaryotic translation initiation factor 4E (eIF-4E) is a 25kDa phosphoprotein that specifically binds to the 7-methylguanosine-containing cap of mRNA. eIF-4E is the rate-limiting component for the initiation of cap-dependent translation by the eIF-4E translation initiation complex. This complex promotes the unwinding of secondary structure at the 5′ untranslated region of mRNA, which is necessary to expose and locate the AUG-initiation codon. Phosphorylation of eIF-4E on Ser-209 occurs after serum treatment in CHO cells, and may regulate its function. Overexpression of eIF-4E can lead to increased cell proliferation, transformation, and tumorigenesis in nude mice. The overexpression of a Ala-53 variant of eIF-4E cannot evoke these changes, suggesting that Ser-53 on eIF-4E participates in the transfer of mRNA to the 48S initiation complexes. In cooperation with nuclear oncogenes such as c-myc or E1A, eIF-4E transforms primary cells. Other studies have demonstrated that overexpression of eIF-4E causes activation of Ras and leads to a transformed phenotype. Subsequent overexpression of GAP then causes reversion of this phenotype. The mechanism by which eIF-4E plays a role in transformation is not clear, but it is postulated that high levels of eIF-4E may lead to the translation of mRNAs that are normally translationally repressed.

Host Species: Mouse
Clone: 87
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Rabbit eIF-4E aa. 1-217

Immunofluorescence, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF-4E
Applications Western Blot
Classification Monoclonal
Clone 87
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Frog, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.