Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eIF-5 Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB611050 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611050 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611050 Supplier BD Biosciences Supplier No. 611050

Mouse Monoclonal Antibody

Initiation of eukaryotic translation involves a series of reactions mediated by multiple eukaryotic initiation factors (eIFs). As a prerequisite to initiation, the initiator methionyl-tRNA (Met-tRNAf), in a ternary complex with eIF-2 and GTP, binds to the 40S ribosomal subunit. This 40S preinitiation complex is positioned at the initiation AUG codon to form the 40S initiation complex. This complex is joined by the 60S ribosomal subunit to form the 80S initiation complex, which functions as a peptidyl transferase. The joining of the 60S subunit to the 40S initiation complex requires eIF-5. This protein associates with the 40S initiation complex, specifically with the β subunit of eIF-2, prior to binding of the 60S subunit. eIF-5 induces GTP hydrolysis of the ternary complex bound to the 40S ribosome. This results in the release of eIF-2 and GDP from the 40S subunit and allows for interaction with the 60S subunit. Furthermore, eIF-5 is thought to be involved in recognition of the start codon and interacts with eIF-3, an additional factor required for 40S initiation complex formation. Thus, eIF-5 is essential to the assembly and proper function of the eukaryotic machinery. eIF-5 has a calculated molecular weight of 49kDa, but has been reported to migrate in a range between 49-53kDa.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen eIF-5
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Host Species Mouse
Immunogen Human eIF-5 aa. 100-215
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.