Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 2A, 65kDa, Mouse, Clone: 3D5, Abnova™

Catalog No. 89014340 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-340 Supplier Abnova Corporation Supplier No. H00083939M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EIF2A.

EIF2A is a 65kDa protein that catalyzes the formation of puromycin-sensitive 80S preinitiation complexes (Zoll et al., 2002 [PubMed 12133843]).[supplied by OMIM

Sequence: MAPSTPLLTVRGSEGLYMVNGPPHFTESTVFPRESGKNCKVCIFSKDGTLFAWGNGEKVNIISVTNKGLLHSFDLLKAVCLEFSPKNTVLATWQPYTTSK

Specifications

Antigen eukaryotic translation initiation factor 2A, 65kDa
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EIF2A.
Formulation PBS with no preservative; pH 7.4
Gene EIF2A
Gene Accession No. NM_032025
Gene Alias CDA02/EIF-2A/MST089/MSTP004/MSTP089
Gene Symbols EIF2A
Host Species Mouse
Immunogen EIF2A (NP_114414, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83939
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.