Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 2-alpha kinase 2, Mouse, Clone: 1D11, Abnova™

Catalog No. 89000908 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-000-908 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-000-908 Supplier Abnova Corporation Supplier No. H00005610M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant EIF2AK2.

Sequence: MAGDLSAGFFMEELNTYRQKQGVVLKYQELPNSGPPHDRRFTFQVIIDGREFPEGEGRSKKEAKNAAAKLAVEILNKEKKAVSPLLLTTTNSSEGLSMGN

Specifications

Antigen eukaryotic translation initiation factor 2-alpha kinase 2
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1D11
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant EIF2AK2.
Formulation PBS with no preservative; pH 7.4
Gene EIF2AK2
Gene Accession No. NM_002759
Gene Alias EIF2AK1/MGC126524/PKR/PRKR
Gene Symbols EIF2AK2
Host Species Mouse
Immunogen EIF2AK2 (NP_002750, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 5610
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.