Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018625 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-625 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-625 Supplier Abnova Corporation Supplier No. H00008892D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human EIF2B2 protein.

Eukaryotic initiation factor-2B (EIF2B) is a GTP exchange protein essential for protein synthesis. It consists of alpha (EIF2B1; MIM 606686), beta (EIF2B2), gamma (EIF2B3; MIM 606273), delta (EIF2B4; MIM 606687), and epsilon (EIF2B5; MIM 603945) subunits. EIF2B activates its EIF2 (see MIM 603907) substrate by exchanging EIF2-bound GDP for GTP.[supplied by OMIM

Sequence: MPGSAAKGSELSERIESFVETLKRGGGPRSSEEMARETLGLLRQIITDHRWSNAGELMELIRREGRRMTAAQPSETTVGNMVRRVLKIIREEYGRLHGRSDESDQQESLHKLLTSGGLNEDFSFHYAQLQSNIIEAINELLVELEGTMENIAAQALEHIHSNEVIMTIGFSRTVEAFLKEAARKRKFHVIVAECAPFCQGHEMAVNLSKAGIETTVMTDAAIFAVMSRVNKVIIGTKTILANGALRAVTGTHTLALAAKHHSTPLIVCAPMFKLSPQFPNEEDSFHKFVAPEEVLPFTEGDILEKVSVHCPVFDYVPPELITLFISNIGGNAPSYIYRLMSELYHPDDHVL

Specifications

Antigen eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human EIF2B2 protein.
Formulation PBS with no preservative; pH 7.4
Gene EIF2B2
Gene Accession No. NM_014239.2
Gene Alias EIF-2Bbeta/EIF2B
Gene Symbols EIF2B2
Host Species Rabbit
Immunogen EIF2B2 (NP_055054.1, 1 a.a. ∼ 351 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8892
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.