Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EIF2B2, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89209719 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-209-719 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-209-719 Supplier Abnova Corporation Supplier No. H00008892K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human EIF2B2 peptide using ARM Technology.

Eukaryotic initiation factor-2B (EIF2B) is a GTP exchange protein essential for protein synthesis. It consists of alpha (EIF2B1; MIM 606686), beta (EIF2B2), gamma (EIF2B3; MIM 606273), delta (EIF2B4; MIM 606687), and epsilon (EIF2B5; MIM 603945) subunits. EIF2B activates its EIF2 (see MIM 603907) substrate by exchanging EIF2-bound GDP for GTP.[supplied by OMIM

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen EIF2B2
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description eukaryotic translation initiation factor 2B, subunit 2 beta, 39kDa
Formulation In 1X PBS, pH 7.4
Gene EIF2B2
Gene Accession No. EIF2B2
Gene Alias EIF-2Bbeta, EIF2B
Gene Symbols EIF2B2
Host Species Rabbit
Immunogen A synthetic peptide of human EIF2B2
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8892
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.